missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 1/CD49a Antibody (CL7207), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Marca: Novus Biologicals NBP2-76478-25ul
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Integrin alpha 1/CD49a Monoclonal specifically detects Integrin alpha 1/CD49a in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated, Block/Neutralize.
Especificaciones
| Integrin alpha 1/CD49a | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| ITGA1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: RQISRSFFSGTQERKVQRN | |
| Primary | |
| Specificity of Integrin alpha 1/CD49a antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Neutralization | |
| CL7207 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Knockdown Validated, Block/Neutralize | |
| CD49 antigen-like family member A, CD49a, CD49a antigen, integrin, alpha 1, Laminin and collagen receptor, very late activation protein 1, VLA-1, VLA1integrin alpha-1 | |
| Mouse | |
| 25 μL | |
| 3672 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido