missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha 1/CD49a Antibody (CL7207), Novus Biologicals™

Código de producto. 18051006 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
25 μL
100 μL
Tamaño de la unidad:
100 microlitros
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18051006 100 μL 100 microlitros
18685181 25 μL 25 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18051006 Supplier Novus Biologicals Supplier No. NBP276478

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 1 publication

Integrin alpha 1/CD49a Monoclonal specifically detects Integrin alpha 1/CD49a in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated, Block/Neutralize.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Integrin alpha 1/CD49a
Aplicaciones Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Neutralization
Clasificación Monoclonal
Clon CL7207
Conjugado Unconjugated
Dilución Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Knockdown Validated, Block/Neutralize
Formulación PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide
Alias de gen CD49 antigen-like family member A, CD49a, CD49a antigen, integrin, alpha 1, Laminin and collagen receptor, very late activation protein 1, VLA-1, VLA1integrin alpha-1
Símbolos de los genes ITGA1
Especie del huésped Mouse
Inmunógeno This antibody was developed against Recombinant Protein corresponding to amino acids: RQISRSFFSGTQERKVQRN
Cantidad 100 μL
Primario o secundario Primary
ID de gen (Entrez) 3672
Especificidad de la prueba Specificity of Integrin alpha 1/CD49a antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.