missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Insulysin/IDE Monoclonal antibody specifically detects Insulysin/IDE in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Especificaciones
Especificaciones
| Antígeno | Insulysin/IDE |
| Aplicaciones | Western Blot, Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | S04-4E8 |
| Concentración | 0.3 mg/mL |
| Conjugado | Unconjugated |
| Formulación | 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Alias de gen | Abeta-degrading protease, EC 3.4.24, EC 3.4.24.56, FLJ35968, Insulin protease, insulinase, insulin-degrading enzyme, Insulysin |
| Especie del huésped | Rabbit |
| Inmunógeno | A synthetic peptide of human Insulysin/IDE (Uniprot # P14735) |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?