missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Mouse, Clone: E2-E3 + 2D11-H5 (same as INS04 + INS05), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-34261-0.1MG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Insulin Monoclonal specifically detects Insulin in Human, Mouse, Porcine, Bovine, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
| Insulin | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Mouse | |
| 6 kDa | |
| 0.1 mg | |
| Diabetes Research, Immune System Diseases, Immunology, Stem Cell Markers | |
| 3630 | |
| Human, Mouse, Pig, Bovine, Rabbit | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| E2-E3 + 2D11-H5 (same as INS04 + INS05) | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/mlimmunohistochemistry-Paraffin 0.5-1ug/ml, SDS-Page | |
| P01308 | |
| INS | |
| Full length (1-84 amino acid) purified pig insulin, conjugated to KLH (E2-E3 and 2D11-H5) | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognizes a polypeptide which is identified as insulin, a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. Deficiency of insulin results in diabetes mellitus. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as beta-cell and insulinoma marker. | |
| Store at 4C. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido