Learn More
iNOS Antibody (4E5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 19 publications
Marca: Novus Biologicals NBP2-22119SS
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
iNOS Monoclonal specifically detects iNOS in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).
Especificaciones
| Monoclonal | |
| iNOS Antibody (4E5) was made to a purified recombinant fragment of human iNOS (C-terminus) expressed in E. coli. [UniProt# P35228] | |
| 0.025 mL | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |