missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Inhibin beta A Antibody (SR4080), Novus Biologicals™
Shop All Bio Techne ProductsDescription
Inhibin beta A Monoclonal antibody specifically detects Inhibin beta A in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antígeno | Inhibin beta A |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | SR4080 |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:2000 |
| Formulación | PBS (pH 7.4), 150mM NaCl, 50% glycerol |
| Alias de gen | Activin beta-A chain, beta A (activin A, activin AB alpha polypeptide), EDF, erythroid differentiation factor, Erythroid differentiation protein, follicle-stimulating hormone-releasing protein, FRP, FSH-releasing protein, inhibin beta A chain, inhibin, beta A, Inhibin, beta-1 |
| Especie del huésped | Rabbit |
| Inmunógeno | A synthesized peptide derived from human Inhibin beta A (Uniprot #: P08476) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?