missing translation for 'onlineSavingsMsg'
Learn More

Influenza A nucleoprotein Antibody (010), Alexa Fluor™ 750, Novus Biologicals™

Código de producto. 18518726 Tienda Bio Techne Productos
Change view
Click to view available options
:
100 μg; Alexa Fluor 750
Product Code.
18518726 100 μg; Alexa Fluor 750
1 options
This item is not returnable. View return policy

Product Code. 18518726

Brand: Novus Biologicals NBP312741AF750

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Influenza A nucleoprotein Monoclonal antibody specifically detects Influenza A nucleoprotein in Influenza A virus H3N2 samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Influenza A nucleoprotein
Aplicaciones Western Blot, ELISA
Clasificación Monoclonal
Clon 010
Conjugado Alexa Fluor 750
Formulación 50mM Sodium Borate
Alias de gen Influenza A virus NP, NP, NP-A, Nucleocapsid protein, Nucleoprotein
Especie del huésped Rabbit
Inmunógeno This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A H3N2 (Uniprot#: P22435.2; Met1-Asn498).
Método de purificación Protein A purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
Especies diana Influenza A virus H3N2
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.