missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin alpha 5/KPNA1/SRP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-76297
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Importin alpha 5/KPNA1/SRP1 Polyclonal antibody specifically detects Importin alpha 5/KPNA1/SRP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Especificaciones
| Importin alpha 5/KPNA1/SRP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| importin subunit alpha-1, importin-alpha-S1, IPOA5, karyopherin alpha 1 (importin alpha 5), Karyopherin subunit alpha-1, NPI-1SRP1importin alpha 5, Nucleoprotein interactor 1, RCH2RAG cohort protein 2, recombination activating gene cohort 2, SRP1-beta | |
| Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human KPNA1. The immunogen is located within amino acids 30 - 80 of KPNA1. Amino Acid Squence: NVATAEEETEEEVMS | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 2.5-20 μg/mL | |
| NP_002255 | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 3836 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido