missing translation for 'onlineSavingsMsg'
Learn More

ILT7/CD85g/LILRA4 Antibody (17G10.2) - Low Endotoxin, Novus Biologicals™

Código de producto. 18669496 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mg
Tamaño de la unidad:
0.10 miligramo
Código de producto. Cantidad unitSize
18669496 0.1 mg 0.10 miligramo
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 18669496

Brand: Novus Biologicals NBP262226

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ILT7/CD85g/LILRA4 Monoclonal antibody specifically detects ILT7/CD85g/LILRA4 in Human samples. It is validated for Flow Cytometry, Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antígeno ILT7/CD85g/LILRA4
Aplicaciones Flow Cytometry, Functional Assay
Clasificación Monoclonal
Clon 17G10.2
Conjugado Unconjugated
Dilución Flow Cytometry 1-5 μg/mL, Functional
Formulación PBS (pH 7.4), 0.2 um filter sterilized
Alias de gen CD85 antigen-like family member G, CD85g, CD85g antigen, ILT-7, ILT7MGC129598, Immunoglobulin-like transcript 7, leukocyte immunoglobulin-like receptor subfamily A member 4, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 4, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member4, MGC129597
Especie del huésped Mouse
Inmunógeno CD85g/ ILT7, a member of leukocyte immunoglobulin-like receptor family expressed on plasmacytoid dendritic cells, but not on myeloid dendritic cells and other peripheral blood leukocytes.
Método de purificación Protein A purified
Cantidad 0.1 mg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 23547
Especies diana Human
Contenido y almacenamiento Store at 4°C. Do not freeze.
Formulario Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.