missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL33 Mouse anti-Human, Clone: 2A9C7, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66235-1-IG-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
IL-33 (Interleukin-33) is a 270 amino acid, highly divergent protein belonging to the IL-1 family with an IL-1-like C-terminal domain. IL-33 is a dual function protein that may function both as a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties. IL-33 binds to and signals through IL1RL1/ST2 and its stimulation recruits MYD88, IRAK1, IRAK4, and TRAF6. IL-33 activates NF-kappaB and MAP kinases, and drives production of TH2-associated cytokines from in vitro polarized TH2 cells. In vivo, IL-33 induces the expression of IL-4, IL-5, and IL-13 and leads to severe pathological changes in mucosal organs. IL-33 is proteolytically converted to a mature form by CASP1 and is highly expressed in high endothelial venules found in tonsils, Peyer′s patches and mesenteric lymph nodes and is almost undetectable in placenta. Prolonged IL-33 treatment of mice led to the development of eosinophilia, splenomegaly, and severe pathological changes in mucosal organs such as lungs, esophagus and small intestine. Recent experiments have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties. Despite its predicted molecular weight, IL-33 will often run at higher molecular weight in SDS-PAGE. Studies have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine, and an intracellular nuclear factor with transcriptional regulatory properties.Especificaciones
| IL33 | |
| Monoclonal | |
| 1.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95760 | |
| IL33 | |
| IL33 Fusion Protein Ag21430 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
| Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
| 2A9C7 | |
| Unconjugated | |
| IL33 | |
| C9orf26, DVS27, IL 1F11, IL 33, IL1F11, IL33, Interleukin 1 family member 11, interleukin 33, NF HEV, NFEHEV, NFHEV, RP11 575C20.2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 90865 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido