missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-6R alpha Antibody (tocilizumab), Alexa Fluor™ 488, Novus Biologicals™
Tienda Bio Techne ProductosDescription
IL-6R alpha Monoclonal antibody specifically detects IL-6R alpha in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
Specifications
| Antígeno | IL-6R alpha |
| Aplicaciones | Flow Cytometry, ELISA, Functional Assay |
| Clasificación | Monoclonal |
| Clon | tocilizumab |
| Conjugado | Alexa Fluor 488 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | CD126, gp80, IL6Q, IL6R, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor |
| Especie del huésped | Human |
| Inmunógeno | IL-6Ra/ CD126 |
| Método de purificación | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?