Descripción
IL-6 Polyclonal specifically detects IL-6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Especificaciones
Especificaciones
| Antígeno | IL-6 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Inmunógeno | Full length human IL6 Recombinant protein. |
| Cantidad | 0.1 mL |
| Especies diana | Mouse, Rat |
For Research Use Only