missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL-3R alpha/CD123 Monoclonal specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
| Antígeno | IL-3R alpha/CD123 |
| Aplicaciones | Western Blot, Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 18N6B12 |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Dilución | Western Blot 10ug/ml, Flow Cytometry 1ug/0.5x10^6cells∼ |
| N.º de referencia del gen | P26951 |
| Alias de gen | CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174 |
| Símbolos de los genes | IL3RA |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?