missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
IL-38/IL-1F10 Monoclonal antibody specifically detects IL-38/IL-1F10 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA
Especificaciones
Especificaciones
| Antígeno | IL-38/IL-1F10 |
| Aplicaciones | Western Blot, Flow Cytometry, ELISA |
| Clasificación | Monoclonal |
| Clon | 798036R |
| Conjugado | PE |
| Dilución | Western Blot, Flow Cytometry, ELISA |
| Formulación | PBS |
| Alias de gen | interleukin 1 family, member 10 (theta), UNQ6119/PRO20041 |
| Especie del huésped | Rat |
| Inmunógeno | E.coli-derived recombinant mouse IL-38/IL-1F10, Met1-Arg152, Accession # Q8R459 |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?