Learn More
IL-24 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12064-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The IL-24 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IL-24. This antibody recognizes human antigen. The IL-24 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Especificaciones
| IL-24 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 11009 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 167 μg/mL | |
| Rabbit | |
| Primary | |
| Human |