Learn More
IL-2 R beta Antibody (TM-b1) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Marca: Novus Biologicals NBP1-43355-0.5mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
IL-2 R beta Monoclonal specifically detects IL-2 R beta in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Block/Neutralize.
Especificaciones
| IL-2 R beta | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Blocking/Neutralizing 50-200 molar excess | |
| CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-2 receptor subunit beta, IL-2R subunit beta, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| 3560 | |
| Store at 4C. Do not freeze. |
| Flow Cytometry, Immunoprecipitation, Blocking Assay | |
| TM-b1 | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| IL2RB | |
| The immunogen for this antibody was Purified CD122. | |
| 0.5 mg | |
| Primary | |
| Mouse | |
| IgG2b |
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido