missing translation for 'onlineSavingsMsg'
Learn More

IL-17B Antibody (B-B58), DyLight 594, Novus Biologicals™

Código de producto. 18583357 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
Código de producto. Cantidad unitSize
18583357 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 18583357

missing translation for 'mfr': Novus Biologicals NBP318115DL594

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-17B Monoclonal antibody specifically detects IL-17B in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antígeno IL-17B
Aplicaciones ELISA
Clasificación Monoclonal
Clon B-B58
Conjugado DyLight 594
Formulación 50mM Sodium Borate
Alias de gen Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20
Especie del huésped Mouse
Inmunógeno Recombinant human IL-17B. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells
Método de purificación Affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Biologically Active Proteins, Cytokine Research
Primario o secundario Primary
ID de gen (Entrez) 27190
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.