missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IL-17/IL-17A Antibody (secukinumab), Novus Biologicals™

Código de producto. 30047802 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30047802 50 μg 50 microgramos
30047958 1 mg 1 miligramo
30045906 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30047802 Proveedor Novus Biologicals™ N.º de proveedor NBP32867850ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Human Monoclonal Antibody

IL-17/IL-17A Monoclonal antibody specifically detects IL-17/IL-17A in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antígeno IL-17/IL-17A
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon secukinumab
Conjugado Unconjugated
Formulación Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
N.º de referencia del gen Q16552
Alias de gen CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
Especie del huésped Human
Inmunógeno CTLA-8/ IL-17a
Método de purificación Protein A purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity
Primario o secundario Primary
ID de gen (Entrez) 3605
Reconstitución Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Especies diana Human
Contenido y almacenamiento Store at -20°C in powder form. Store at -80°C once reconstituted.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.