missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
IL-12/IL-23 p40 Monoclonal antibody specifically detects IL-12/IL-23 p40 in Mouse samples. It is validated for ELISA
Especificaciones
Specifications
| Antígeno | IL-12/IL-23 p40 |
| Aplicaciones | ELISA |
| Clasificación | Monoclonal |
| Clon | 240 |
| Conjugado | Alexa Fluor 350 |
| Dilución | ELISA |
| Formulación | 50mM Sodium Borate |
| Alias de gen | CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40 |
| Especie del huésped | Rabbit |
| Inmunógeno | Rabbit immunized with purified, recombinant Mouse IL-12 p40/IL12B (Uniprot#: P43432, Met1-Ser335) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?