missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
IL-12 Monoclonal antibody specifically detects IL-12 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Functional Assay, Radioimmune Assays
Especificaciones
Especificaciones
| Antígeno | IL-12 |
| Aplicaciones | Western Blot, ELISA, Immunohistochemistry, Functional Assay, Radioimmune Assays (RIA) |
| Clasificación | Monoclonal |
| Clon | 1-1D5 |
| Conjugado | mFluor Violet 450 SE |
| Dilución | Western Blot, ELISA, Immunohistochemistry, Functional, Radioimmunoassay |
| Formulación | 50mM Sodium Borate |
| Alias de gen | CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 |
| Especie del huésped | Mouse |
| Inmunógeno | Recombinant human IL-12 produced by baculovirus. |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?