Learn More
IKZF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 12016-1-AP-150UL
 Ver más versiones de este producto
Código de producto. 16849473
  • 410.00€ / 150 microlitros
Descripción
RNP2 a type I integral membrane protein found only in the rough endoplasmic reticulum. The encoded protein is part of an N-oligosaccharyl transferase complex that links high mannose oligosaccharides to asparagine residues found in the Asn-X-Ser/Thr consensus motif of nascent polypeptide chains. This protein is similar in sequence to the yeast oligosaccharyl transferase subunit SWP1.
Especificaciones
Especificaciones
IKZF1
Polyclonal
Unconjugated
IKZF1
DNA binding protein Ikaros, hIk 1, Hs.54452, IK1, IKAROS, IKZF1, LYF1, PRO0758, ZNFN1A1
Rabbit
Antigen Affinity Chromatography
RUO
10320, 22778, 305501
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q03267, Q13422
IKZF1
IKZF1 Fusion Protein Ag2631
150 μL
Primary
Human, Mouse, Rat