missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IHH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13388-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a member of the hedgehog family of secreted signaling molecules. Hedgehog proteins are essential regulators of a variety of developmental processes including growth, patterning and morphogenesis. The encoded protein specifically plays a role in bone growth an differentiation. Mutations in this gene are the cause of brachydactyly type A1 which is characterized by shortening or malformation of the phalanges. Mutations in this gene are also the cause of acrocapitofemoral dysplasia.Especificaciones
| IHH | |
| Polyclonal | |
| Unconjugated | |
| IHH | |
| BDA1, HHG 2, HHG2, IHH, Indian hedgehog protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16147, 3549, 84399 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P97812, Q14623 | |
| IHH | |
| IHH Fusion Protein Ag4046 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido