missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
IgG4 Monoclonal antibody specifically detects IgG4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ELISA, ELISA
Especificaciones
Especificaciones
| Antígeno | IgG4 |
| Aplicaciones | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | RM120 |
| Conjugado | Unconjugated |
| Dilución | Western Blot : 0.5 ug/ml - 2 ug/ml, Flow Cytometry : 1ug/ml-4ug/ml, ELISA, Immunohistochemistry : 1 - 10 ug/ml, Immunocytochemistry/ Immunofluorescence : 1 - 10 ug/ml, Immunohistochemistry-Paraffin : 1 - 10 ug/ml, Sandwich ELISA Capture : 50 - 200ng/well, Sandwich ELISA Detection : 0.05 - 0.2ug/ml |
| Formulación | PBS |
| Alias de gen | Immunoglobulin G, Immunoglobulin G4, ImmunoglobulinG |
| Especie del huésped | Rabbit |
| Inmunógeno | Peptide corresponding to the hinge region of Human IgG4 |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?