missing translation for 'onlineSavingsMsg'
Learn More

IGF-I R/IGF1R Antibody - BSA Free, Novus Biologicals™

Product Code. 18218756 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.025 mL
0.1 mL
Unit Size:
0.10 ml
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18218756 0.025 mL 25 microlitros
18706493 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18218756 Supplier Novus Biologicals Supplier No. NBP177680SS

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 2 publications

IGF-I R/IGF1R Polyclonal specifically detects IGF-I R/IGF1R in Human, Mouse, Rat, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno IGF-I R
Aplicaciones Western Blot, Immunocytochemistry, Immunofluorescence
Clasificación Polyclonal
Concentración 1.09 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 1:50-1:100
Formulación PBS and 30% Glycerol with 0.05% Sodium Azide
N.º de referencia del gen P08069
Alias de gen CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF-I receptor, IGFIR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2
Símbolos de los genes IGF1R
Especie del huésped Rabbit
Inmunógeno A synthetic peptide made to an internal portion of the human IGF1 Receptor protein (between residues 700-800) [UniProt P08069]
Método de purificación Affinity Purified
Cantidad 0.025 mL
Estado normativo RUO
Disciplina de investigación Apoptosis, Cancer, Lipid and Metabolism, Neuroscience, Nutrient Sensing in the Brain, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primario o secundario Primary
ID de gen (Entrez) 3480
Especificidad de la prueba Human, rat, mouse and primate. Reactivity with Equine reported via customer feedback. Predicted to react with Bovine (100%).
Especies diana Human, Mouse, Rat, Equine, Primate
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.