missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgE Monoclonal Antibody (Ige21), NovaFluor™ Yellow 730, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen H077T02Y07
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
IgE is one of the five classes of immunoglobulins found in mammals. Monomers consist of two heavy (epsilon) chains with 4 Ig-like constant domains, and two light chains. IgE plays an essential role in type I hypersensitivity (allergic responses such as hay fever, asthma, hives, anaphylatic shock) by binding to Fc receptors on basophils and mast cells. Crosslinking of IgE bound to Fc receptors induces degranulation of mast cells and basophils that promote allergic manifestations. IgE is also functionally important for immunity against parasites.
Especificaciones
| IgE | |
| Monoclonal | |
| 0.2 μg/Test | |
| PBS with 0.09% sodium azide, pH 17.2 | |
| P01854 | |
| IGHE | |
| Affinity chromatography | |
| RUO | |
| 3497 | |
| 4°, C, store in dark, DO NOT FREEZE! | |
| IgG1 κ |
| Flow Cytometry | |
| Ige21 | |
| NovaFluor Yellow 730 | |
| IGHE | |
| HCAK1, IgA1, IgG3, IGHA1, IGHA2, IGHG1, IGHG2, IGHG3, IGHG4, IGKC, IGKCD, IGLC, IGLC1, Immunoglobulin E, immunoglobulin heavy constant alpha 1, immunoglobulin heavy constant alpha 2 (A2m marker), immunoglobulin heavy constant gamma 1 (G1m marker), immunoglobulin heavy constant gamma 2 (G2m marker), immunoglobulin heavy constant gamma 3 (G3m marker), immunoglobulin heavy constant gamma 4 (G4m marker), immunoglobulin kappa constant, immunoglobulin lambda constant 1, ImmunoglobulinE, Km | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido