missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
IFN-gamma R1/CD119 Monoclonal antibody specifically detects IFN-gamma R1/CD119 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | IFN-gamma R1/CD119 |
| Aplicaciones | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | JE40-50 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:2000 |
| Formulación | 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol |
| Alias de gen | AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFN-gamma-R1, IFNGR, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2 |
| Especie del huésped | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?