Learn More
IDH2 Mouse, anti-Human, CoraLite Plus 488, Clone: 1F10A10, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-66918-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The IDH2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IDH2. This antibody recognizes human antigen. The IDH2 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Especificaciones
| IDH2 | |
| 1F10A10 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG1 |
| Western Blot | |
| 1000 μg/mL | |
| ICD M, IDH, IDH2, IDHM, IDP, IDPM, mNADP IDH, NADP(+) specific ICDH, Oxalosuccinate decarboxylase | |
| 20 μL | |
| 3418 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |