missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10403-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is a rabbit polyclonal antibody raised against a full-length human ICT1 protein.
Essential peptidyl-tRNA hydrolase component of the mitochondrial large ribosomal subunit. Acts as a codon-independent translation release factor that has lost all stop codon specificity and directs the termination of translation in mitochondrion, possibly in case of abortive elongation. May be involved in the hydrolysis of peptidyl-tRNAs that have been prematurely terminated and thus in the recycling of stalled mitochondrial ribosomes.Especificaciones
| ICT1 | |
| Polyclonal | |
| Unconjugated | |
| MRPL58 | |
| Digestion substraction 1, DS 1, DS1, ICT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 303673, 3396, 68572 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14197, Q8R035 | |
| MRPL58 | |
| ICT1 Fusion Protein Ag0647 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido