missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-2 Recombinant Rabbit Monoclonal Antibody (033), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen MA543107
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Specificity: Human ICAM-2/CD102
ICAM-2 is a member of the intercellular adhesion molecule (ICAM) family. All ICAM proteins are type I transmembrane glycoproteins, contain 2-9 immunoglobulin-like C2-type domains, and bind to the leukocyte adhesion LFA-1 protein. ICAM-2 may play a role in lymphocyte recirculation by blocking LFA-1-dependent cell adhesion. It mediates adhesive interactions important for antigen-specific immune response, NK-cell mediated clearance, lymphocyte recirculation, and other cellular interactions important for immune response and surveillance. Several transcript variants encoding the same protein have been found for this gene. Diseases associated with ICAM2 include Colon Carcinoma In Situ.
Especificaciones
ICAM-2 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P13598 | |
ICAM2 | |
Recombinant Human ICAM-2/CD102 protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
033 | |
Unconjugated | |
ICAM2 | |
CD102; Icam2; ICAM-2; Intercellular adhesion molecule 2; Ly-60; Lymphocyte function-associated AG-1 counter-receptor; RP23-418O11.10-001 | |
Rabbit | |
Protein A | |
RUO | |
3384 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
ICAM-2 Recombinant Rabbit Monoclonal Antibody (033), Invitrogen™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido