Learn More
HVCN1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14162-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
HVCN1 (hydrogen voltage-gated channel 1), a voltage gated proton channel protein that is highly expressed in immune tissues, mediates the proton conductances required by phagocytic leukocytes for the oxidative burst that underlies microbial killing. HVCN1 moderates the voltage-dependent proton permeability of excitable membranes by allowing the flow of protons in accordance to their electrochemical gradient. HVCN1 is sensitive to zinc ions, and can be inhibited by them. HVCN1 may represent a new therapeutic target for B cell maligcies on continued signaling via the B cell antigen receptor.Especificaciones
| HVCN1 | |
| Polyclonal | |
| Unconjugated | |
| HVCN1 | |
| HV1, HVCN1, VSOP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84329 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96D96 | |
| HVCN1 | |
| HVCN1 Fusion Protein Ag5350 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |