missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HUNK, GST Tag Recombinant Protein

Código de producto. 17168745
Change view
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
Código de producto. Cantidad unitSize
17168745 100 μg 100 microgramos
15920303 10 μg 10 microgramos
17118795 1 mg 1 miligramo
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17168745

Marca: Invitrogen™ A32892

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This HUNK recombinant human protein (full length) was expressed in insect cells. HUNK recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for HUNK is NP_055401.1.

HUNK serine/threonine kinase or hormonally up-regulated Neu-associated kinase is a member of the mitogen-activated protein kinase family which is essential for mammary tumor metastasis. HUNK deletion in mice did not alter viability, fertility, longevity, or propensity to develop tumors and it is also dispensable for murine development. HUNK has been shown to participate in vesicular transport. Additionally, functional studies in mice suggest that HUNK regulates endocytosis via interaction with rabaptin-5 protein and induces changes in mammary glands during pregnancy. HUNK expression has been documented in mouse brain, breast, embryo, and fetus. ESTs have been isolated from several human tissue libraries, including normal ear.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P57058
Concentración See Label
Para utilizar con (aplicación) Kinase Assay
Formulación 50mM tris HCl with 0.25mM DTT, 150mM NaCl, 0.1mM PMSF, 10mM glutathione, 25% glycerol, 0.1mM EDTA and no preservative; pH 7.5
ID de gen (Entrez) 30811
Peso molecular 105.9 kDa
Nombre Human HUNK, GST Tag
Intervalo de pH 7.5
Método de purificación Purified
Cantidad 100 μg
Requisitos de almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Estado normativo RUO
Alias de gen B19; Bstk1; Hormonally up-regulated neu tumor-associated kinase; hormonally upregulated neu tumor-associated kinase; hormonally up-regulated Neu-associated kinase; hormonally upregulated Neu-associated kinase; Hunk; MAKV; Mak-v; RGD1565091; serine/threonine protein kinase MAK-V; serine/threonine-protein kinase MAK-V
Nombre común HUNK
Símbolo de gen HUNK
Familia Quinasa CAMKL Kinase Family
Tipo de producto Protein
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína GST-tag
Secuencia Full length
Contenido y almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Condiciones de envío Dry Ice
Sistema de expresión Insect cells
Familia de proteínas Kinases & Inhibitors
Forma de proteína Full Length, Recombinant, Active
Formulario Liquid
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.