missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Número de acceso | Q9HBE4 |
| Formulación | 1 x PBS, See Certificate of Analysis for details |
| ID de gen (Entrez) | 59067 |
| Peso molecular | 18 kDa, monomer, glycosylated |
| Cantidad | 10 μg |
| Fuente | HEK293 |
| Requisitos de almacenamiento | Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.) |
| Concentración de endotoxinas | <1 EU/μg |
| Reactividad específica | Human |
| Recombinante | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?