missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
DKK-1 (Dickkopf WNT signaling pathway inhibitor 1) is a member of the DKK protein family. Other members of this family include DKK-2, DKK-3, and DKK-4. The DKK family proteins share 35-40 % homology between various members. Recombinant human DKK-1 expressed in HEK293 cells is a 35-45kDa glycosylated protein. It is a secreted protein that functions as a negative regulator of Wnt/β-catenin signaling pathway. Wnt/ β-catenin signaling pathway involves binding of the Wnt ligands to the seven-transmembrane receptor, Frizzled, and the coreceptor lipoprotein-related protein 5 and 6 (LRP5/6). DKK1 forms a ternary complex with LRP5/6 and another receptor, Kremen, followed by endocytosis of this complex and removal of LRP5/6 from the cell surface. It has been suggested that DKK-1 permits anterior development by inhibiting Wnt/ β-catenin signaling, which is essential for posterior patterning in vertebrates. The balance between Wnt signaling and DKK-1 inhibition is critical for bone formation and homeostasis and insufficient or excess DKK-1 activity in bone results in increased or decreased bone density, respectively. (PMID: 17143291; 12050670; 10570958)
Especificaciones
Especificaciones
| Número de acceso | O94907 |
| Formulación | 1 x PBS, See Certificate of Analysis for details |
| ID de gen (Entrez) | 22943 |
| Peso molecular | 35-38 kDa |
| Cantidad | 100 μg |
| Fuente | HEK293 |
| Requisitos de almacenamiento | Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.) |
| Concentración de endotoxinas | <1 EU/μg |
| Reactividad específica | Human |
| Recombinante | Recombinant |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?