missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human TNF-alpha Recombinant Protein

Código de producto. 10699920
Change view
Click to view available options
Cantidad:
10 μg
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
10699920 10 μg cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 10699920

Marca: Gibco™ PHC3015L

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Carrier-Free Storage: Store the liquid human TNF-alpha at ≤-20°C. Upon thawing, apportion into working aliquots and store at ≤-20°C. Avoid repeated freeze-thaw cycles.

TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P01375
Para utilizar con (aplicación) Bioactivity
Formulación 40mM tris with no preservative; pH 8
ID de gen (Entrez) 7124
Peso molecular 17.5 kDa
Nombre Human TNF-alpha
Intervalo de pH 8
Método de purificación Purified
Cantidad 10 μg
Requisitos de almacenamiento -20°C
Estado normativo RUO
Concentración de endotoxinas <0.1 ng/μg
Alias de gen APC1 protein; Cachectin; C-domain 1; C-domain 2; cTNF; DADB-70P7.1; DIF; ICD1; ICD2; Intracellular domain 1; Intracellular domain 2; N-terminal fragment; NTF; RATTNF; Tnf; TNF alpha; TNF superfamily; TNF α; TNF, macrophage-derived; TNF, monocyte-derived; TNFA; TNF-a; TNFalpha; TNF-alpha; Tnfsf1a; TNFSF2; TNFα; TNLG1F; Tumor necrosis factor; tumor necrosis factor (TNF superfamily, member 2); tumor necrosis factor alpha; tumor necrosis factor alpha (cachetin); tumor necrosis factor alpha precursor; tumor necrosis factor ligand 1F; tumor necrosis factor ligand superfamily member 2; Tumor necrosis factor, membrane form; Tumor necrosis factor, soluble form; tumor necrosis factor-alpha; tumor necrosis factor-alpha precursor; tumor-necrosis factor; tumour necrosis factor
Nombre común TNF alpha
Símbolo de gen Tnf
Actividad biológica ED50 = 0.02 - 0.05 ng/mL; determined by the dose-dependent cytotoxcity on L929 cells in the presence of actinomycin D.
Tipo de producto Protein
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Contenido y almacenamiento -20°C
Actividad ED50 = 0.02 - 0.05 ng/mL; determined by the dose-dependent cytotoxcity on L929 cells in the presence of actinomycin D.
Nivel de endotoxinas <0.1 ng/μg
Condiciones de envío Dry Ice
Sistema de expresión E. coli
Familia de proteínas Cytokines & Receptors
Forma de proteína Recombinant, Ligand
Formulario Liquid
Subtipo de proteína TNF (Tumor Necrosis Factor)
Categoría de investigación Immunology, Signal Transduction, Stem Cell Research, Inflammation, Oncology
Línea de productos Gibco
Grado de pureza o calidad >95% by SDS-PAGE
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.