missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TMA16 (aa 44-119) Control Fragment Recombinant Protein

Código de producto. 30199743
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30199743

Marca: Invitrogen™ RP106410

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111360 (PA5-111360. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TMA16 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso Q96EY4
Concentración ≥5.0 mg/mL
Para utilizar con (aplicación) Blocking Assay, Control
Formulación 1 M urea, PBS with no preservative; pH 7.4
ID de gen (Entrez) 55319
Nombre Human TMA16 (aa 44-119) Control Fragment
Cantidad 100 μL
Estado normativo RUO
Alias de gen 1810029B16Rik; C4orf43; RGD1305222; TMA16; translation machinery associated 16 homolog; translation machinery associated 16 homolog (S. cerevisiae); translation machinery-associated protein 16; UPF0534 protein C4orf43; UPF0534 protein C4orf43 homolog
Nombre común TMA16
Símbolo de gen TMA16
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína His-ABP-tag
Secuencia KALRLNLVGEKLQWFQNHLDPQKKRYSKKDACELIERYLNRFSSELEQIELHNSIRDRQGRRHCSRETVIKQTMER
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Sistema de expresión E. coli
Formulario Liquid
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.