missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human TGF-alpha Recombinant Protein, PeproTech®
Click to view available options
Cantidad:
100 μg
1 mg
20 μg
250 μg
500 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
250 microgramos
500 microgramos
Descripción
Recombinant human TGF-alpha is a 50 amino acid polypeptide (5.5 kDa) which shares approximately 40% sequence homology with EGF, including 6 conserved cysteine residues, which form 3 intramolecular disulfide bonds. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
TGF-alpha is an EGF-related polypeptide growth factor that signals through the EGF receptor, and stimulates the proliferation of a wide range of epidermal and epithelial cells. It is produced by monocytes, keratinocytes, and various tumor cells. TGF-alpha induces anchorage-independence transformation in cultured cells. Human, murine and rat TGF-alpha are cross-species reactive. TGFalpha (aa 50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.
Especificaciones
Especificaciones
| Número de acceso | P01135 |
| Para utilizar con (aplicación) | ELISA, Functional Assay, Western Blot |
| Formulación | protein with no preservative |
| ID de gen (Entrez) | 7039 |
| Peso molecular | 5.5 kDa |
| Nombre | Human TGF-alpha |
| Cantidad | 1 mg |
| Fuente | E. coli |
| Estado normativo | RUO |
| Concentración de endotoxinas | <1 EU/ μg |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido