missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SSC5D (aa 870-962) Control Fragment Recombinant Protein

Código de producto. 30181200
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
30181200 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30181200

Marca: Invitrogen™ RP99201

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59819 (PA5-59819. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Binds to extracellular matrix proteins. Binds to pathogen-associated molecular patterns (PAMPs) present on the cell walls of Gram-positive and Gram-negative bacteria and fungi, behaving as a pattern recognition receptor (PRR). Induces bacterial and fungal aggregation and subsequent inhibition of PAMP-induced cytokine release. Does not possess intrinsic bactericidal activity. May play a role in the innate defense and homeostasis of certain epithelial surfaces.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso A1L4H1
Concentración ≥5.0 mg/mL
Para utilizar con (aplicación) Blocking Assay, Control
Formulación 1 M urea, PBS with no preservative; pH 7.4
ID de gen (Entrez) 284297
Nombre Human SSC5D (aa 870-962) Control Fragment
Cantidad 100 μL
Estado normativo RUO
Alias de gen S5D-SRCRB; scavenger receptor cysteine rich domain containing (5 domains); scavenger receptor cysteine rich family member with 5 domains; scavenger receptor cysteine rich family, 5 domains; scavenger receptor cysteine-rich glycoprotein; soluble scavenger protein with 5 SRCR domains; Soluble scavenger receptor cysteine-rich domain-containing protein SSC5D; soluble scavenger with 5 domains; SSC5D
Nombre común SSC5D
Símbolo de gen SSC5D
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína His-ABP-tag
Secuencia TCTGYTDYDDYPPWTWDPTSREDLAKGTTTAGVPGHTLPWRTTRRPGSSSPAIRRLPDTGSKDGYKLPWTWDTPSGRGLAEGTPTAGKLGPTL
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Sistema de expresión E. coli
Formulario Liquid
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.