Learn More
STEMCELL Technologies Human Recombinant SCF
Descripción
- Stem cell factor (SCF) is an early-acting cytokine that plays a pivotal role in the regulation of embryonic and adult hematopoiesis
- SCF promotes cell survival, proliferation, differentiation, adhesion, and functional activation of cells at multiple levels of the hematopoietic hierarchy
- Together with other cytokines such as thrombopoietin and Flt3/Flk-2 Ligand, SCF is commonly used to promote expansion of primitive hematopoietic stem cells and multipotent progenitor cells in culture
- In synergy with various growth factors, including IL-2, IL-3, IL-6, IL-7, G-CSF, and erythropoietin, SCF increases proliferation and differentiation of myeloid and erythroid progenitor cells and a subset of lymphoid progenitor cells
- SCF is also a primary growth and activation factor for mast cells and eosinophils
Especificaciones
Especificaciones
| Nombre | Kit ligand, Mast cell growth factor, Steel factor, Stem cell factor |
| Cantidad | 100 μg |
| Tipo de célula | Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Pluripotent Stem Cells |
| Grado de pureza o calidad | ≥95% |