Learn More
STEMCELL Technologies Human Recombinant R-Spondin-1
Descripción
- R-Spondin-1 (RSPO1) is the prototype member of the R-Spondin (RSPO) protein subfamily of a superfamily of thrombospondin type 1 repeat (TSR-1)-containing proteins
- Although unable to initialize signaling, RSPO family members are potent enhancers of WNT signaling
- They are characterized by a TSR-1 domain, a carboxy-terminal region with positively charged amino acids, and two N-terminal furin-like cysteine-rich repeats
- R-Spondin-1 activates β-catenin signaling via the WNT signaling cascade and by indirectly increasing low-density lipoprotein receptor-related protein (LRP6) on the cell surface
Especificaciones
Especificaciones
| Nombre | hRspo1, Roof plate-specific Spondin-1 |
| Cantidad | 25 μg |
| Tipo de célula | Endoderm, PSC-Derived; Epithelial Cells; Hematopoietic Stem and Progenitor Cells; Intestinal Cells; Mesoderm, PSC-Derived; Other; Pluripotent Stem Cells |
| Grado de pureza o calidad | ≥95% |