Learn More
STEMCELL Technologies Human Recombinant PDGF-AA
Código de producto. 17199551
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1000 μg
100 μg
10 μg
Tamaño de la unidad:
10 microgramos
100 microgramos
1000 microgramos
17199551 1000 μg 1000 microgramos
17169551 10 μg 10 microgramos
17179551 100 μg 100 microgramos
3 options
Código de producto. 17199551
Marca: STEMCELL Technologies 78095.3
Platelet-derived growth factor AA
  • Platelet-derived growth factor (PDGF) is a dimeric glycoprotein consisting of two disulfide bridge-stabilized polypeptide chains, A and B, which are assembled as heterodimers (PDGF-AB) or homodimers (PDGF-AA and PDGF-BB)
  • PDGF signals through the receptor tyrosine kinases PDGFRalpha and PDGFRbeta
  • It has been shown that PDGF-induced migration involves signaling pathways involving MEK/ERK, EGFR, Src, and PI3K/AKT
  • PDGF is a potent mitogen for cells of mesenchymal origin, such as fibroblasts, glial cells, and vascular smooth muscle cells
  • PDGF has been implicated in pathogenesis of atherosclerosis, glomerulonephritis, cancer, and in the contraction of vascular smooth muscle cells of rat aortic tissues
  • It has been suggested that PDGF-AA is an important autocrine regulator of vascular endothelial growth factor (VEGF) expression in non-small cell lung carcinomas
Especificaciones
Cantidad 1000 μg
Tipo de célula Mesenchymal Cells, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Other
Grado de pureza o calidad ≥95%