Learn More
STEMCELL Technologies Human Recombinant Noggin
Click to view available options
Cantidad:
1000 μg
10 μg
50 μg
Tamaño de la unidad:
10 microgramos
1000 microgramos
50 microgramos
Descripción
- Noggin binds to and antagonizes bone morphogenetic protein (BMP) ligands of the transforming growth factor beta (TGF-β) family
- Noggin supports maintenance of undifferentiated human embryonic stem cells in vitro, and can be used to prevent spontaneous differentiation in the short term (Chaturvedi et al.) Noggin is essential for development of structures derived from ectoderm embryonic somite, skeletal patterning, and neurogenesis in vivo
- It also influences chondrogenesis, osteogenesis, and joint formation (Krause et al.), and promotes dopaminergic differentiation of embryonic stem cells and subsequent survival of dopamine neurons (Chiba et al.)
Especificaciones
Especificaciones
| Nombre | NOG, SYM1, SYNS1, Symphalangism 1 (proximal), Synostoses (multiple) syndrome |
| Cantidad | 50 μg |
| Tipo de célula | Neural Cells, PSC-Derived; Neurons; Pluripotent Stem Cells |
| Grado de pureza o calidad | ≥95% |