Learn More
STEMCELL Technologies Human Recombinant Myostatin
Descripción
- Myostatin is a member of transforming growth factor beta (TGF-β) superfamily that signals by binding to activin type II receptor, resulting in the recruitment of either ALK3 or ALK4 coreceptor, and activation of SMAD2/3 (Kondás et al.) It is released by myocytes and inhibits muscle growth and ability to regenerate (Lee & Lee)
- Myostatin appears to affect the lipid catabolic metabolism of adipocytes and inhibits preadipocyte differentiation, as demonstrated in the 3T3-L1 cell line (Li et al.)
Especificaciones
Especificaciones
| Nombre | GDF-8, Growth differentiation factor 8, MSTN78 |
| Cantidad | 100 μg |
| Tipo de célula | Mesenchymal Stem and Progenitor Cells |
| Grado de pureza o calidad | ≥95% |