Learn More
STEMCELL Technologies Human Recombinant MIP-1 beta (CCL4)
Click to view available options
Cantidad:
25 μg
5 μg
Tamaño de la unidad:
25 microgramos
5 microgramos
Descripción
- Macrophage inflammatory protein-1 beta (MIP-1 beta), also known as CCL4, is a member of the CC family of chemokines and is most closely related to CCL3 or MIP-1 alpha
- MIP-1 beta is predominantly synthesized by activated CD8+ T cells, specifically the perforin low memory CD8+ T cells
- MIP-1 beta signals through CCR5, which is the major coreceptor required for entry of certain strains of HIV-1 into permissive cells, and it is known that HIV-1-specific CD8+ T cell clones that exhibit cytotoxic T lymphocyte (CTL) activity secrete MIP-1 beta upon encountering sensitized target cells
- MIP-1 beta attracts a variety of immune cells to sites of microbial infection
- In vitro experiments show that human MIP-1 beta tends to attract CD4+ T lymphocytes, preferentially of the naïve (CD45RA) phenotype
Especificaciones
Especificaciones
| Nombre | ACT-2, Immune activation protein 2, LAG-1, Lymphocyte activation gene 1 protein, MIP-1b, Protein H400, SCYA2, SCYA4, Small-inducible cytokine A4, T-cell activation protein 2 |
| Cantidad | 25 μg |
| Tipo de célula | B Cells; Dendritic Cells; Mesenchymal Stem and Progenitor Cells; Monocytes; NK Cells; Other; T Cells; T Cells, CD4+; T Cells, CD8+ |
| Grado de pureza o calidad | ≥95% |