Learn More
STEMCELL Technologies Human Recombinant M-CSF
Código de producto. 17159461
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1000 μg
100 μg
10 μg
Tamaño de la unidad:
100 microgramos
1000 microgramos
17159461 100 μg 100 microgramos
16244551 10 μg 100 microgramos
17169461 1000 μg 1000 microgramos
3 options
Código de producto. 17159461
Marca: STEMCELL Technologies 78057
Macrophage colony-stimulating factor
  • Macrophage colony-stimulating factor (M-CSF) is a homodimeric glycoprotein growth factor that regulates proliferation and differentiation of myeloid hematopoietic progenitor cells to mononuclear phagocytic cell lineages, including monocytes, macrophages, and osteoclasts
  • M-CSF is a crucial factor for the development of tissue-resident macrophages in most tissues
  • It is required for the maturation and activation of monocytes and macrophages, and regulates inflammatory responses in conjunction with other stimuli such as IFN-γ, LPS, and IL-4
  • M-CSF is also required for bone resorption by osteoclasts, and is involved in the development and regulation of the placenta, mammary gland, and brain
  • M-CSF is produced by monocytes, fibroblasts, osteoclasts, stromal cells, endothelial cells, and tumor cells
Especificaciones
Nombre Colony stimulating factor 1, CSF-1
Cantidad 100 μg
Tipo de célula Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Monocytes; Myeloid Cells; Neurons
Grado de pureza o calidad ≥95%