Learn More
STEMCELL Technologies Human Recombinant IL-7, ACF
Código de producto. 17119811
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1000 μg
100 μg
10 μg
Tamaño de la unidad:
10 microgramos
100 microgramos
1000 microgramos
17119811 100 μg 100 microgramos
17109811 10 μg 10 microgramos
17129811 1000 μg 1000 microgramos
3 options
Código de producto. 17119811
Marca: STEMCELL Technologies 78196.1
Interleukin 7, animal component-free
  • Interleukin 7 (IL-7) is a member of the type I cytokine family that is critical for T and B cell development and survival
  • It is produced by non-hematopoietic cells in the thymus, lymphoid organs, and by bone marrow stromal cells (Lundström et al.) IL-7 binds to a receptor (IL-7R) composed of common gamma chain and IL-7Ra (CD127) and signals through the JAK/STAT and PI3K pathways
  • IL-7 regulates the survival of naïve and memory CD4+ and CD8+ T cells, γδ T cells, NK T cells, innate lymphoid cells, and regulatory T cells (Carrette & Surh)
  • Although a deficiency in IL-7R still permits the generation of normal numbers of peripheral B cells in humans, stimulation of human B cell precursors with IL-7 could promote STAT5-dependent proliferation and survival in vitro (Clark et al.; Corfe & Paige)
  • This product is animal component-free
Especificaciones
Nombre Interleukin-7, LP-1, Lymphopoietin 1, Pre-B cell factor
Cantidad 100 μg
Tipo de célula Hematopoietic Stem and Progenitor Cells
Grado de pureza o calidad ≥95%