Learn More
STEMCELL Technologies Human Recombinant IL-6R alpha
Código de producto. 17129531
Encompass_Preferred
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
50 microgramos
Product Code. 17129531
missing translation for 'mfr': STEMCELL Technologies 78083.1
This item is not returnable. View return policy
Interleukin 6 receptor alpha
  • Interleukin 6 receptor (IL-6R) alpha is a type I transmembrane glycoprotein that forms a complex with type I transmembrane signal transducer protein gp130 (CD130) and mediates the biological activities of IL-6
  • IL-6 binds to the membrane-bound non-signaling IL-6R alpha (mIL-6R), and the complex binds to two molecules of gp130 and leads to ‘classical' IL-6-signal transduction, which includes activation of JAK/STAT, ERK, and PI3K signal transduction pathways
  • In contrast, a soluble form of IL-6R alpha (sIL-6R), which comprises the extracellular portion of the receptor, binds to the secreted IL-6 to form a complex that promotes bioavailability of IL-6
  • The complex of IL-6 and sIL-6R can bind to gp130 on cells that do not express the IL-6R and are unresponsive to IL-6
  • This process is known as trans-signaling
Specifications
Nombre B cell stimulatory factor-2, CD126, IL-6R, IL-6R1, IL-6RA, Interleukin-6 receptor, Interleukin-6 receptor alpha
Cantidad 50 μg
Grado de pureza o calidad ≥95%
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.