Learn More
STEMCELL Technologies Human Recombinant GDF-11
Click to view available options
Cantidad:
100 μg
20 μg
Tamaño de la unidad:
100 microgramos
20 microgramos
Descripción
- Growth differentiation factor 11 (GDF-11) is a member of transforming growth factor beta (TGF-β) family
- It binds to the TGF-β receptors ALK4, ALK5, and ALK7 and activates the SMAD signaling pathway (Ho et al.) GDF-11 regulates the development of the olfactory system, retina, and pancreas, as well as anterior/posterior patterning of the axial skeleton (Lee & Lee)
- GDF-11 is an endocrine factor expressed in skeletal muscle, brain, and dental pulp (Kondás et al.) Studies in mice showed that GDF-11 regulates muscle and cardiac aging, and stimulates neurogenesis by remodeling blood vessels (Katsimpardi et al.; Loffredo et al.; Sinha et al.)
Especificaciones
Especificaciones
| Nombre | BMP-11, Bone morphogenetic protein 11, Growth differentiation factor 11 |
| Cantidad | 100 μg |
| Tipo de célula | Neural Cells, PSC-Derived |
| Grado de pureza o calidad | ≥95% |