Learn More
STEMCELL Technologies Human Recombinant FGF-8B, ACF
Código de producto. p-7217398
Encompass_Preferred
Click to view available options
Cantidad:
1000 μg
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
1000 microgramos
25 microgramos
Código de producto. 17109831
Marca: STEMCELL Technologies 78204
Fibroblast growth factor 8B, animal component-free
  • Fibroblast growth factor 8B (FGF-8B) is a member of the fibroblast growth factor (FGF) family and is an isoform of FGF-8
  • Cytokines in the FGF family possess broad mitogenic and cell survival activities (Folkman & Klagsbrun; Kimelman & Kirschner) and are involved in a variety of biological processes, including cell proliferation, differentiation, survival, and apoptosis (Folkman & Klagsbrun; Klagsbrun; Rifkin & Moscatelli)
  • FGF-8B signals through FGF receptors (FGFRs) to activate PI3K and MAPK pathways
  • FGF-8B regulates gastrulation, epithelial-mesenchymal transition, and mesenchymal to epithelial differentiation during embryonic development
  • FGF-8B has also been found in peripheral blood leukocytes and healthy bone marrow samples (Mattila & Härkönen)
  • FGF-8B has mitogenic effects on somatic cells in the germinal epithelium and is expressed in adult mouse ovarian cells and tissues, which suggests that it regulates maturation of oocytes and seminiferous epithelium in testes (Valve et al.) This product is animal component-free
Especificaciones
Nombre AIGF, Androgen-induced growth factor, FGF-8, Fibroblast growth factor-8B, HBGF-8, Heparin-binding growth factor-8
Cantidad 25 μg
Tipo de célula Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Neural Cells, PSC-Derived; Pluripotent Stem Cells
Grado de pureza o calidad ≥95%
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.