Learn More
STEMCELL Technologies Human Recombinant FGF-8B, ACF
Click to view available options
Cantidad:
1000 μg
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
1000 microgramos
25 microgramos
Descripción
- Fibroblast growth factor 8B (FGF-8B) is a member of the fibroblast growth factor (FGF) family and is an isoform of FGF-8
- Cytokines in the FGF family possess broad mitogenic and cell survival activities (Folkman & Klagsbrun; Kimelman & Kirschner) and are involved in a variety of biological processes, including cell proliferation, differentiation, survival, and apoptosis (Folkman & Klagsbrun; Klagsbrun; Rifkin & Moscatelli)
- FGF-8B signals through FGF receptors (FGFRs) to activate PI3K and MAPK pathways
- FGF-8B regulates gastrulation, epithelial-mesenchymal transition, and mesenchymal to epithelial differentiation during embryonic development
- FGF-8B has also been found in peripheral blood leukocytes and healthy bone marrow samples (Mattila & Härkönen)
- FGF-8B has mitogenic effects on somatic cells in the germinal epithelium and is expressed in adult mouse ovarian cells and tissues, which suggests that it regulates maturation of oocytes and seminiferous epithelium in testes (Valve et al.) This product is animal component-free
Especificaciones
Especificaciones
| Nombre | AIGF, Androgen-induced growth factor, FGF-8, Fibroblast growth factor-8B, HBGF-8, Heparin-binding growth factor-8 |
| Cantidad | 25 μg |
| Tipo de célula | Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Neural Cells, PSC-Derived; Pluripotent Stem Cells |
| Grado de pureza o calidad | ≥95% |