Learn More
STEMCELL Technologies Human Recombinant FGF-8B
Descripción
- Fibroblast growth factor 8B (FGF-8B) is a member of the fibroblast growth factor (FGF) family and is an isoform of FGF-8
- Cytokines in the FGF family possess broad mitogenic and cell survival activities and are involved in a variety of biological processes, including cell proliferation, differentiation, survival, and apoptosis
- FGF-8B signals through FGF receptors (FGFRs) to activate PI3K and MAPK pathways
- FGF-8B is broadly associated with mitogenic and cell survival activities, and regulates gastrulation, epithelial-mesenchymal transition, and later on mesenchymal to epithelial differentiation during embryonic development
- FGF-8B has also been found in peripheral blood leukocytes and healthy bone marrow samples
Especificaciones
Especificaciones
| Nombre | AIGF, Androgen-induced growth factor, FGF-8, FGF-8B, Fibroblast growth factor-8B, HBGF-8, Heparin-binding growth factor 8; HH6, KAL6 |
| Cantidad | 1000 μg |
| Tipo de célula | Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Neural Cells, PSC-Derived; Pluripotent Stem Cells |
| Grado de pureza o calidad | >95% |