Learn More
STEMCELL Technologies Human Recombinant FGF-8B
Código de producto. 17179331
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1000 μg
50 μg
Tamaño de la unidad:
1000 microgramos
50 microgramos
17179331 1000 μg 1000 microgramos
17169331 50 μg 50 microgramos
2 options
Código de producto. 17179331
Marca: STEMCELL Technologies 78008.2
Fibroblast growth factor 8B
  • Fibroblast growth factor 8B (FGF-8B) is a member of the fibroblast growth factor (FGF) family and is an isoform of FGF-8
  • Cytokines in the FGF family possess broad mitogenic and cell survival activities and are involved in a variety of biological processes, including cell proliferation, differentiation, survival, and apoptosis
  • FGF-8B signals through FGF receptors (FGFRs) to activate PI3K and MAPK pathways
  • FGF-8B is broadly associated with mitogenic and cell survival activities, and regulates gastrulation, epithelial-mesenchymal transition, and later on mesenchymal to epithelial differentiation during embryonic development
  • FGF-8B has also been found in peripheral blood leukocytes and healthy bone marrow samples
Especificaciones
Nombre AIGF, Androgen-induced growth factor, FGF-8, FGF-8B, Fibroblast growth factor-8B, HBGF-8, Heparin-binding growth factor 8; HH6, KAL6
Cantidad 1000 μg
Tipo de célula Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Neural Cells, PSC-Derived; Pluripotent Stem Cells
Grado de pureza o calidad >95%