Learn More
STEMCELL Technologies Human Recombinant FGF-7 (KGF)
Código de producto. 17169431
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1000 μg
10 μg
50 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
50 microgramos
17169431 50 μg 50 microgramos
15887502 1000 μg 1 miligramo
17159431 10 μg 10 microgramos
3 options
Código de producto. 17169431
Marca: STEMCELL Technologies 78046.1
Fibroblast growth factor 7
  • Fibroblast growth factor 7 (FGF-7) is a member of the FGF family, and acts exclusively through a subset of FGF receptor isoforms expressed predominantly by epithelial cells (Finch & Rubin)
  • FGF-7 seems to act specifically on epithelial cells and stimulates proliferation, migration, and differentiation of these cells, and also participates in epithelial protection and repair both in vitro and in vivo (Finch & Rubin; Werner)
  • In contrast, FGF-7 is produced solely by cells of mesenchymal origin, and functions as a paracrine mediator of mesenchymal-epithelial communication (Rubin et al.) FGF-7 has also been shown to supplement several wound-healing properties of bioengineered skin (Erdag et al.) and to induce autophagy in human keratinocytes (Belleudi et al.) Additionally, FGF-7 has a role in pluripotent stem cell differentiation to endodermal pancreatic-like insulin-producing cells and thymic epithelial cells (Inami et al.; Niu et al.)
Especificaciones
Nombre Fibroblast growth factor-7, HBGF-7, Heparin-binding growth factor 7, Keratinocyte growth factor
Cantidad 50 μg
Tipo de célula Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Keratinocytes; Mammary Cells; Mesenchymal Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells; Prostate Cells
Grado de pureza o calidad ≥95%